Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Violet 540)

Lieferant: AAT BIOQUEST
10280120 10280121
AATB10280120EA 371 CHF
AATB10280120 AATB10280121
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Violet 540)
Antikörper
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR