Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Violet 510)

Lieferant: AAT BIOQUEST
11610110 11610111
AATB11610110EA 371 CHF
AATB11610110 AATB11610111
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Violet 510)
Antikörper
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR