Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (APC (Allophycocyanin))
Lieferant: AAT BIOQUEST
116101C0 116101C1 116101C2
AATB116101C0EA 271 CHF
AATB116101C0 AATB116101C1 AATB116101C2
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (APC (Allophycocyanin))
Antikörper
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human