Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (TRITC)

Lieferant: AAT BIOQUEST
103801I0 103801I1
AATB103801I0EA 371 CHF
AATB103801I0 AATB103801I1
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (TRITC)
Antikörper
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: TRITC
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR