Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PacBlue)
Lieferant: AAT Bioquest
AATB101411J0EA 371 CHF
AATB101411J0 AATB101411J1
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PacBlue)
Antikörper
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: PacBlue
Epitope:
Host: Mouse
Isotype:
Reactivity: Human