Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 750)
Lieferant: AAT Bioquest
AATB100410L1EA 1390 CHF
AATB100410L1 AATB100410L0
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (iFluor® 750)
Antikörper
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: iFluor® 750
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human