Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Violet 510)

Lieferant: AAT BIOQUEST
10081110 10081111
AATB10081110EA 371 CHF
AATB10081110 AATB10081111
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (mFluor™ Violet 510)
Antikörper
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR