Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Violet 500)
Lieferant: AAT BIOQUEST
10280100 10280101
AATB10280100EA 371 CHF
AATB10280100 AATB10280101
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (mFluor™ Violet 500)
Antikörper
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
  • Affinity purified
  • Easily reconstituted
Bitte beachten For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human