Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE (Phycoerythrin)/Cy5®)

Lieferant: AAT BIOQUEST
104411M0 104411M1 104411M2
AATB104411M0EA 319 CHF
AATB104411M0 AATB104411M1 AATB104411M2
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE (Phycoerythrin)/Cy5®)
Antikörper
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Bitte beachten For research use only.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR