Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (Pacific Orange)
Lieferant: AAT BIOQUEST
104411K0 104411K1
AATB104411K0EA 371 CHF
AATB104411K0 AATB104411K1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (Pacific Orange)
Antikörper
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: Pacific Orange
Epitope:
Host: Rat
Isotype:
Reactivity: Human