Human Recombinant Repulsive guidance molecule A (from E. coli)
Lieferant: ProSci
PRSI92-257EA 0 CHF
PRSI92-257
Human Recombinant Repulsive guidance molecule A (from E. coli)
Proteine und Peptide
Repulsive guidance molecule A(RGMA) is a cell membrane protein and belongs to the repulsive guidance molecule (RGM) family. It interacts with NEO1, BMP2 and BMP4. RGMA is a glycosylphosphatidylinositol-anchored glycoprotein that functions as an axon guidance protein in the developing and adult central nervous system. It helps guide Retinal Ganglion Cell (RGC) axons to the tectum in the midbrain. RGMa has been implicated to play an important role in the developing brain and in the scar tissue that forms after a brain injury. This protein may also function as a tumor suppressor in some cancers.
  • Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays.
Bitte beachten For research use only.