Anti-Rabbit IgG Goat Polyclonal Antibody (CF®750)
Lieferant: Biotium
20919-50UL 20919-250UL
20919-50ULEA 123 CHF
20919-50UL 20919-250UL
Anti-Rabbit IgG Goat Polyclonal Antibody (CF®750)
Antikörper
Goat anti-rabbit IgG (H+L), F(ab')2 fragment secondary antibody labeled with our superior CF® Dyes and other labels.
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues.
Bitte beachten Storage of the antibody for more than a day at final working dilution is not recommended.