Riboflavin (Vitamin B2)
Lieferant: Chemicon
Synonyms:
Lactoflavin
, Vitamin B2
, 6,7-Dimethyl-9-D-ribitylisoalloxazin
, 7,8-Dimethyl-10-(D-ribo-2,3,4,5-tetrahydroxypentyl)-benzo[g]pteridin-2,4(3H,10H)-dion
, Vitamin G
, (-)-Riboflavin
CHMCAB5773EA
0
CHF
CHMCAB5773
Riboflavin (Vitamin B2)
Riboflavin (Vitamin B2)
Formel:
C₁₇H₂₀N₄O₆ MW: 376,37 g/mol Melting Pt: 280 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00005022 CAS-Nummer: 83-88-5 EINECS: 201-507-1 Merck Index: 12,08367 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...