Triamteren

Lieferant: LGC Standards PROMOCHEM

PROOMM0269.00EA 2100 CHF
PROOMM0269.00
Triamteren
Triamteren
Formel: C₁₂H₁₁N₇
MW: 253,27 g/mol
Siedepunkt: 573 °C (1013 hPa)
Melting Pt: 316,1 °C
MDL Number: MFCD00006708
CAS-Nummer: 396-01-0
EINECS: 206-904-3
Merck Index: 12,09731

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR