Eriochromschwarz T

Lieferant: Roth Carl
Achtung

Synonyms: Natrium-3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalinsulfonat1-(2-Hydroxy-1-naphthylazo)-6-nitro-2-naphthol-4-sulfonsäure Natriumsalz

ROTHK860.1EA 51.35 CHF
ROTHK860.1
Eriochromschwarz T
Eriochromschwarz T
Formel: C₂₀H₁₂N₃NaO₇S
MW: 461,39 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003935
CAS-Nummer: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III
Merck Index: 13,03702

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR