Bromphenolblau Natriumsalz

Lieferant: EUROCLONE

EMR105005 EMR105010 EMR105025
EURLEMR105005EA 0 CHF
EURLEMR105005 EURLEMR105010 EURLEMR105025
Bromphenolblau Natriumsalz
Bromphenolblau Natriumsalz
Formel: C₁₉H₉Br₄NaO₅S
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013793
CAS-Nummer: 34725-61-6
EINECS: 252-170-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR