Kongorot
Lieferant: Molekula
|
Gefahr
|
Synonyms:
Direct Red 28
, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalin-1-sulfonat)
, Direktrot 28
MOLE24120761-100GEA
0
CHF
MOLE24120761-100G
Kongorot
Kongorot
Formel:
C₃₂H₂₂N₆Na₂O₆S₂ MW: 696,67 g/mol Melting Pt: > 360 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00004028 CAS-Nummer: 573-58-0 EINECS: 209-358-4 Merck Index: 12,02562 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...