Colchicin

Lieferant: LATOXAN
Gefahr

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

L6051-1 L6051-200
LATOL6051-1EA 0 CHF
LATOL6051-1 LATOL6051-200
Colchicin
Colchicin
Formel: C₂₂H₂₅NO₆
MW: 399,44 g/mol
Melting Pt: 154…156 °C
Dichte: 1.32 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00078484
CAS-Nummer: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1,I
Merck Index: 12,02536

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR