Anti-Human IgG Goat Polyclonal Antibody (CF®583R)

Lieferant: Biotium
20901-50UL 20901-500UL
20901-50ULEA 123 CHF
20901-50UL 20901-500UL
Anti-Human IgG Goat Polyclonal Antibody (CF®583R)
Antikörper
Highly cross-adsorbed goat anti-human IgG (H L) secondary antibody labeled with our superior CF® dyes.

  • Highly cross-adsorbed for specific staining with minimal background
  • Available in 11 bright and photostable CF® dyes
  • HRP and R-PE conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

This is a highly cross-adsorbed goat anti-human IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against bovine, horse, and mouse serum.

Type: Secondary
Antigen: Human IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR