5-Fluoruracil
Lieferant: ENZO LIFE SCIENCES AG
Synonyms:
2,4-Dihydroxy-5-fluorpyrimidin
, Fluoruracil
, 5-FU
ENZOALX480099G005EA
137
CHF
ENZOALX480099G005
5-Fluoruracil
5-Fluoruracil
Thymidylate synthetase inhibitor
Potent antitumor agent. Inhibits thymidylate synthetase. Induces p53-dependent apoptosis.
Formel:
C₄H₃FN₂O₂ MW: 130,08 g/mol |
MDL Number:
MFCD00006018 CAS-Nummer: 51-21-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...