Cyclopiazonic acid ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES AG

ENZOBMLCA4150050EA 728 CHF
ENZOBMLCA4150050
Cyclopiazonic acid ≥98% (durch TLC)
Cyclopiazonic acid

ATPase inhibitor


Mycotoxin. Cell permeable. Induces the release of intracellular stored Ca²⁺ without increasing IP3 levels via inhibition of endoplasmic reticulum Ca²⁺-ATPase. Highly specific inhibitor of Ca²⁺-ATPase of sarcoplasmic reticulum, completely inhibiting the enzyme at 6-8 nmol/mg protein (at 0.5-2.0µM ATP).

Formel: C₂₀H₂₀N₂O₃
MW: 336,391 g/mol
MDL Number: MFCD00167445
CAS-Nummer: 18172-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR