Puromycin-Aminonukleosid ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES AG

ENZOBMLA2600050EA 317 CHF
ENZOBMLA2600050
Puromycin-Aminonukleosid ≥98% (durch TLC)
Puromycin-Aminonukleosid

A puromycin analog which does not inhibit protein synthesis or induce apoptosis. It is a glomerular epithelial cell toxin which may be used to induce nephropathy in laboratory animals.


  • White solid


Soluble in water (10 mg/ml)


Verpackung: Shipping temperature: Ambient

Formel: C₁₂H₁₈N₆O₃
MW: 294,31 g/mol
Melting Pt: 222…223 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00063462
CAS-Nummer: 58-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR