Mycophenolsäure ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES AG
ENZOBMLA2490500EA
421
CHF
ENZOBMLA2490500
Mycophenolsäure ≥98% (durch TLC)
Mycophenolsäure
Inosine-5'-monophosphate dehydrogenase inhibitor
Antiviral and immunosuppressive activity. Clinically useful antipsoriatic agent. Mycophenolic acid prodrugs are effective in rheumatoid arthritis and for the prevention of organ transplant rejection. Mechanism of action is inhibition of inosine-5'-monophosphate dehydrogenase.
Isolated from Penicillium brevi-compactum
Formel:
C₁₇H₂₀O₆ MW: 320,34 g/mol |
MDL Number:
MFCD00036814 CAS-Nummer: 24280-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...