Acetylcholinchlorid

Lieferant: ENZO LIFE SCIENCES AG

Synonyms: (2-Acetoxyethyl)trimethylammoniumchloridAcetylcholiniumchlorid

ENZOALX550135G005EA 59.2 CHF
ENZOALX550135G005
Acetylcholinchlorid
Acetylcholinchlorid
Formel: H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
MW: 181,66 g/mol
Melting Pt: 151 °C
Dichte: 1,263 g/cm³ (20 °C)
MDL Number: MFCD00011698
CAS-Nummer: 60-31-1
EINECS: 200-468-8
Merck Index: 13,00089

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR