Muscimol ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES AG
|
Gefahr
|
ENZOALX550127M005EA
197
CHF
ENZOALX550127M005
Muscimol ≥98% (durch TLC)
Muscimol
Potent GABAA receptor agonist.
Very soluble in water (10 mg/ml) or methanol; soluble in 100% ethanol.
Formel:
C₄H₆N₂O₂ Lager Temperatur: Gefrierschrank |
MDL Number:
MFCD00057894 CAS-Nummer: 2763-96-4 EINECS: 220-430-4 UN: 1544 ADR: 6.1,II Merck Index: 14,06312 |
Spezifikation Testergebnisse
Purity (TLC) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...