Tamoxifen citrate ≥99%
Lieferant: ENZO LIFE SCIENCES AG
ENZOALX550095G001EA
226
CHF
ENZOALX550095G001
Tamoxifen citrate ≥99%
Tamoxifen citrate
PKC inhibitor
Antiestrogen with anti-cancer activity. Selective protein kinase C (PKC) inhibitor and induces PKC epsilon translocation. Induces apoptosis in estrogen receptor-negative human cancer cell lines.
Formel:
C₂₆H₂₉NO MW: 563,65 g/mol |
MDL Number:
MFCD00058321 CAS-Nummer: 54965-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...