Natriumorthovanadat
Lieferant: ENZO LIFE SCIENCES AG
|
Achtung
|
ENZOALX400032G005EA
60.5
CHF
ENZOALX400032G005
Natriumorthovanadat
Natriumorthovanadat
Formel:
Na₃O₄V MW: 183,91 g/mol Melting Pt: 850…866 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003511 CAS-Nummer: 13721-39-6 EINECS: 237-287-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...