7-Oxostaurosporine (aus Streptomyces sp. MST-AS5345) ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES AG

ENZOALX380210M001EA 716 CHF
ENZOALX380210M001
7-Oxostaurosporine (aus Streptomyces sp. MST-AS5345) ≥98% (durch HPLC)
7-Oxostaurosporine

Antibiotic. Potent inhibitor of protein kinase C (PKC). Inhibits the cell cycle at the G2 stage.


  • Apoptosis inducer. PKC inhibitor.


Yellow solid.


Soluble in DMSO, dimethyl formamide, methanol or 100% ethanol; slightly soluble in water.

Formel: C₂₈H₂₄N₄O₄
MW: 480,5 g/mol
Lager Temperatur: Gefrierschrank
CAS-Nummer: 125035-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR