Anti-Mouse IgG Chicken Polyclonal Antibody (CF®583R)

Lieferant: Biotium
20890-500UL 20890-50UL
20890-500ULEA 248 CHF
20890-500UL 20890-50UL
Anti-Mouse IgG Chicken Polyclonal Antibody (CF®583R)
Antikörper
This is a chicken anti-mouse IgG (H+L) secondary antibody labeled with our superior CF® Dyes.

  • Available in seven bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Conjugates of blue fluorescent dyes like CF®350 are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colours.

Type: Secondary
Antigen: Mouse IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Chicken
Isotype: IgG (H+L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR