Kongorot ≥95% (durch HPLC), ultrarein
Lieferant: ENZO LIFE SCIENCES AG
Synonyms: Direct Red 28Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalin-1-sulfonat)Direktrot 28
ENZOENZ52552EA 158 CHF
ENZOENZ52552
Kongorot ≥95% (durch HPLC), ultrarein
Kongorot
beta-Amyloid dye
Congo Red, has been used in microscopy for staining elastic fibers and bacteria. Amyloid protein, an abnormal protein aggregate associated with various pathologies, is commonly detected in tissue with Congo Red. Detection of amyloid is relevant for a whole host of diseases, including Alzheimer’s disease, Creutzfeld-Jakob’s disease and Bovine Spongiform Encephalopathy. The birefringence of Congo Red-stained amyloid, appearing green under polarized light, improves detection relative to simple visible examination of the colored deposits. Additionally, upon binding to amyloid, Congo Red displays bright fluorescence emission at 614 nm. Wavelength Maxima: Excitation 497 nm.
Formel: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696,67 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00004028
CAS-Nummer: 573-58-0
https://ch.vwr.com/stibo/web/std.lang.all/16/25/45401625.jpg