Ölsäure ≥99% (durch GC)

Lieferant: ENZO LIFE SCIENCES AG

Synonyms: cis-9-Octadecensäure

ENZOBMLFA0450010EA 136 CHF
ENZOBMLFA0450010
Ölsäure ≥99% (durch GC)
Ölsäure

PKC activator


Major mono-unsaturated fatty acid constituent of olive oil occurring as the glyceride. Activates protein kinase C (PKC) in hepatocytes. Inhibitor of glucose production and food intake. Upregulates the expression of uncoupling protein-3 (UCP-3) in L6 myotubes. Promotes differentiation of cultured neuronal cells mediated via PPARβ and it induces a myofibroblast-like phenotype in mesangial cells.

Formel: C₁₈H₃₄O₂
MW: 282,47 g/mol
MDL Number: MFCD00064242
CAS-Nummer: 112-80-1
https://ch.vwr.com/stibo/web/std.lang.all/38/56/45403856.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR