β-Alanin
Lieferant: Chemicon
Synonyms:
H-Bala-OH
, H-β-Ala-OH
, 3-Aminopropansäure
, 3-Aminopropionsäure
CHMCAB140EA
0
CHF
CHMCAB140
β-Alanin
β-Alanin
Formel:
C₃H₇NO₂ MW: 89,09 g/mol Siedepunkt: 201 °C (1013 hPa) Melting Pt: 94 °C Dichte: 1,44 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00008200 CAS-Nummer: 107-95-9 EINECS: 203-536-5 Merck Index: 13,00204 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...