L-Alanin
Lieferant: BACHEM BIOCHEMICA
Synonyms:
(S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
4030174.0100
4030174.0250
BACE4030174.0100EA
0
CHF
BACE4030174.0100
BACE4030174.0250
L-Alanin
L-Alanin
Formel:
C₃H₇NO₂ Siedepunkt: 189 °C (1013 hPa) Melting Pt: 79 °C Dichte: 1.4 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00064410 CAS-Nummer: 56-41-7 EINECS: 200-273-8 Merck Index: 13,00203 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...