D-α-Phenylalanin
Lieferant: Molekula
Synonyms:
3-Phenyl-D(+)-alanin
, 3-Phenyl-D-alanin
, H-D-Phe-OH
, (R)-(+)-2-Amino-3-phenylpropionsäure
, (R)-(+)-2-Amino-3-phenylpropansäure
, (R)-(+)-2-Amino-3-phenylpropionic acid
, D-Phenylalanin
MOLEM76521605EA
0
CHF
MOLEM76521605
D-α-Phenylalanin
D-α-Phenylalanin
Formel:
C₉H₁₁NO₂ MW: 165,19 g/mol Siedepunkt: 295 °C (1013 hPa) |
MDL Number:
MFCD00004270 CAS-Nummer: 673-06-3 EINECS: 211-603-5 Merck Index: 12,07425 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...