D-Alanin

Lieferant: Chemicon

Synonyms: α-AlaninD-α-Alanin(R)-(-)-2-AminopropansäureD(-)-2-Aminopropionic acid(R)-(-)-2-AminopropionsäureAlaninD(-)-Alanin(R)-(-)-Alanin(R)-(-)-2-Aminopropionic acidH-D-Ala-OH

CHMCAB5777EA 0 CHF
CHMCAB5777
D-Alanin
D-Alanin
Formel: C₃H₇NO₂
MW: 89,09 g/mol
Siedepunkt: 189 °C (1013 hPa)
Melting Pt: 79 °C
MDL Number: MFCD00008077
CAS-Nummer: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR