Molekularsieb A5 (0,5 nm, 5 Å) Moisture Trap volume 200 cc, 1/8 in. fitting, Supelco®
Lieferant: Merck
20619.EA 253 CHF
20619.
Molekularsieb A5 (0,5 nm, 5 Å) Moisture Trap volume 200 cc, 1/8 in. fitting, Supelco®
Molekularsieb A5 (0,5 nm, 5 Å)
MDL Number: MFCD00131613
CAS-Nummer: 69912-79-4