Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 405)
Lieferant: AAT BIOQUEST
10280020 10280021
AATB10280020EA 371 CHF
AATB10280020 AATB10280021
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 405)
Antikörper
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 405
Epitope:
Host: Mouse
Isotype:
Reactivity: Human