Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (FITC)
Lieferant: AAT BIOQUEST
100521H0 100521H1
AATB100521H0EA 371 CHF
AATB100521H0 AATB100521H1
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (FITC)
Antikörper
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Bitte beachten For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: FITC
Epitope:
Host: Mouse
Isotype:
Reactivity: Human