N-Methyl-N-(trimethylsilyl)trifluoracetamid, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyms:
N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoracetamid
, MSTFA
M-Clarity™ quality level = MQ100
44156-5ML-FEA
309
CHF
44156-5ML-F
N-Methyl-N-(trimethylsilyl)trifluoracetamid, Supelco®
N-Methyl-N-(trimethylsilyl)trifluoracetamid
Formel:
C₆H₁₂F₃NOSi MW: 199,25 g/mol Siedepunkt: 130 °C (1013 hPa) Dichte: 1,07 g/cm³ (20 °C) |
MDL Number:
MFCD00000411 CAS-Nummer: 24589-78-4 EINECS: 246-331-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...