Getreideöl

Lieferant: Roth Carl

Synonyms: Maisöl

ROTH6212.1EA 61.75 CHF
ROTH6212.1
Getreideöl
Getreideöl
Melting Pt: –10 °C
Dichte: 0,918 g/cm³ (20 °C)
MDL Number: MFCD00130865
CAS-Nummer: 8001-30-7
EINECS: 232-281-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR