Clofenotan

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyms: 1,1,1-Trichlor-2,2-bis(4-chlorphenyl)ethanDichlordiphenyltrichlorethan4,4'-DDTDDT1-Chlor-4-(1-(4-chlorphenyl)-2,2,2-trichlorethyl)benzolp,p'-DDTDicophan2,2,2,p,p'-Pentachlorethylidenbisbenzol

PROONMIAP1309EA 452 CHF
PROONMIAP1309
Clofenotan
Clofenotan
Formel: C₁₄H₉Cl₅
MW: 354,49 g/mol
Siedepunkt: 260 °C (1013 hPa)
Melting Pt: ∼89 °C
Dichte: ∼1,5 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
CAS-Nummer: 50-29-3
EINECS: 200-024-3
UN: 2761
ADR: 6.1,III
Merck Index: 12,02898

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR