5-Chloruracil

Lieferant: Molekula

Synonyms: 5-Chlor-2,4-dihydroxypyrimidinChloruracil

MOLEM87447286EA 0 CHF
MOLEM87447286
5-Chloruracil
5-Chloruracil
Formel: C₄H₃ClN₂O₂
MW: 146,53 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD00006019
CAS-Nummer: 1820-81-1
EINECS: 217-339-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR