Platin(IV)oxid

Lieferant: Molekula
Achtung

Synonyms: PlatindioxidAdams Katalysator

MOLEM86173878EA 0 CHF
MOLEM86173878
Platin(IV)oxid
Platin(IV)oxid
Formel: O₂Pt
MW: 227,08 g/mol
Melting Pt: 450 °C
Dichte: 11,8 g/cm³ (20 °C)
MDL Number: MFCD00011184
CAS-Nummer: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II
Merck Index: 12,07686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR