Benzyltributylammoniumchlorid

Lieferant: Molekula
Achtung

MOLE46358428-500GEA 0 CHF
MOLE46358428-500G
Benzyltributylammoniumchlorid
Benzyltributylammoniumchlorid
Formel: C₁₉H₃₄ClN
MW: 311,94 g/mol
Melting Pt: 155…159 °C
MDL Number: MFCD00011849
CAS-Nummer: 23616-79-7
EINECS: 245-787-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR