4-Brombutyronitril

Lieferant: Molekula

Synonyms: 4-Brombuttersäurenitril

MOLEM41593590EA 0 CHF
MOLEM41593590
4-Brombutyronitril
4-Brombutyronitril
Formel: BrH₂CCH₂CH₂CN
MW: 148 g/mol
Siedepunkt: 205 °C (1013 hPa)
Dichte: 1,49 g/cm³ (20 °C)
Flash-Pt: 103 °C
MDL Number: MFCD00001971
CAS-Nummer: 5332-06-9
EINECS: 226-236-6
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR