Allyltrimethylsilan

Lieferant: Molekula

MOLEM36010201EA 0 CHF
MOLEM36010201
Allyltrimethylsilan
Allyltrimethylsilan
Formel: H₂CCHCH₂Si(CH₃)₃
MW: 114,26 g/mol
Siedepunkt: 84 °C (1013 hPa)
Dichte: 0,7139 g/cm³ (20 °C)
MDL Number: MFCD00008635
CAS-Nummer: 762-72-1
EINECS: 212-104-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR