2-Amino-6-methoxybenzothiazol

Lieferant: Molekula

MOLE72628801-25GEA 0 CHF
MOLE72628801-25G
2-Amino-6-methoxybenzothiazol
2-Amino-6-methoxybenzothiazole
Formel: C₈H₈N₂OS
MW: 180,23 g/mol
Siedepunkt: 348 °C (1013 hPa)
Melting Pt: 160 °C
MDL Number: MFCD00005787
CAS-Nummer: 1747-60-0
EINECS: 217-130-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR