Methylundecanoat

Lieferant: MATREYA

Synonyms: Undecansäuremethylester

MAYA1166EA 0 CHF
MAYA1166 BTRN1166
Methylundecanoat
Methylundecanoat
Formel: C₁₂H₂₄O₂
MW: 200,32 g/mol
MDL Number: MFCD00008957
CAS-Nummer: 1731-86-8
EINECS: 217-053-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR