Fentinhydroxid, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyms:
Triphenylzinnhydroxid
M-Clarity™ Qualitätsstufe = MQ100
45493-250MGEA
50.7
CHF
45493-250MG
Fentinhydroxid, Supelco®
Fentinhydroxid
Formel:
C₁₈H₁₆OSn MW: 367,03 g/mol Melting Pt: 128 °C Dichte: 1,54 g/cm³ (20 °C) |
MDL Number:
MFCD00013928 CAS-Nummer: 76-87-9 EINECS: 200-990-6 UN: 3146 ADR: 6.1,III Merck Index: 12,09875 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...