Tricin
Lieferant: EUROCLONE
Synonyms:
N-(Tris(hydroxymethyl)methyl)glycin
EURLEMR297100EA
0
CHF
EURLEMR297100
Tricin
Tricin
Formel:
(HOCH₂)₃CNHCH₂CO₂H MW: 179,17 g/mol Melting Pt: 187 °C Dichte: 1,523 g/cm³ (19.85 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00004277 CAS-Nummer: 5704-04-1 EINECS: 227-193-6 Merck Index: 13,09724 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...